siRNA Details
| Gene Symbol: LOC100506725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108082.1 | 1 | 282 | |||
| NR_108083.1 | 1 | 282 | ||||
| GenBank mRNA | DT218025.1 | - | 1 | 273 | ||
| HY153176.1 | - | 1 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506725| Gene Symbol: LOC100506725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108082.1 | 1 | 282 | |||
| NR_108083.1 | 1 | 282 | ||||
| GenBank mRNA | DT218025.1 | - | 1 | 273 | ||
| HY153176.1 | - | 1 | 283 | |||