siRNA Details
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 1 | 167 | |||
GenBank mRNA | AL832059.1 | - | 1 | 103 | ||
BC092503.1 | - | 1 | 112 | |||
DA100871.1 | - | 1 | 167 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 1 | 167 | |||
GenBank mRNA | AL832059.1 | - | 1 | 103 | ||
BC092503.1 | - | 1 | 112 | |||
DA100871.1 | - | 1 | 167 |