siRNA Details
| Gene Symbol: LOC100132217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128836.1 | - | 3 | 3466 | ||
| Gene Symbol: LOC107986805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012932.1 | XP_016868421.1 | 1 | 1682 | ||

LOC100132217
Genomic Map