siRNA Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516608.2 | XP_011514910.1 | 7, 8 | 977 | ||
| GenBank mRNA | AI674788.1 | - | 1, 2 | 424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IMMP2L| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516608.2 | XP_011514910.1 | 7, 8 | 977 | ||
| GenBank mRNA | AI674788.1 | - | 1, 2 | 424 | ||