siRNA Details
| Gene Symbol: NAPEPLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017011856.1 | XP_016867345.1 | 1 | 33 | ||
| XM_017011857.1 | XP_016867346.1 | 1 | 33 | |||
| GenBank mRNA | AK304644.1 | - | 1 | 33 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAPEPLD| Gene Symbol: NAPEPLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017011856.1 | XP_016867345.1 | 1 | 33 | ||
| XM_017011857.1 | XP_016867346.1 | 1 | 33 | |||
| GenBank mRNA | AK304644.1 | - | 1 | 33 | ||