siRNA Details
| Gene Symbol: KPRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025231.1 | NP_001020402.1 | 2 | 901 | ||
| XM_011509571.1 | XP_011507873.1 | 2 | 942 | |||
| GenBank mRNA | AY960854.1 | - | 1 | 872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KPRP| Gene Symbol: KPRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025231.1 | NP_001020402.1 | 2 | 901 | ||
| XM_011509571.1 | XP_011507873.1 | 2 | 942 | |||
| GenBank mRNA | AY960854.1 | - | 1 | 872 | ||