siRNA Details
| Gene Symbol: SUMO1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002190.1 | 1 | 980 | |||
| GenBank mRNA | AA021599.1 | - | 1 | 222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUMO1P3| Gene Symbol: SUMO1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002190.1 | 1 | 980 | |||
| GenBank mRNA | AA021599.1 | - | 1 | 222 | ||