siRNA Details
| Gene Symbol: TRIM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008274.3 | NP_001008275.2 | 4, 5 | 991 | ||
| XM_017023254.1 | XP_016878743.1 | 3, 4 | 527 | |||
| GenBank mRNA | AK090695.1 | - | 3, 4 | 436 | ||
| AK131485.1 | - | 4, 5 | 936 | |||
| BC033211.1 | - | 4, 5 | 921 | 4906353 | ||
| DA058421.1 | - | 3, 4 | 436 | |||

TRIM72
Genomic Map