siRNA Details
| Gene Symbol: SPANXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009614.2 | NP_001009614.1 | 2 | 397 | ||
| GenBank mRNA | DQ336118.1 | - | 2 | 397 | ||
| DQ336119.1 | - | 2 | 397 | |||
| DQ336120.1 | - | 2 | 397 | |||
| DQ336121.1 | - | 2 | 397 | |||

SPANXN1
Genomic Map