siRNA Details
| Gene Symbol: CHAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008708.2 | NP_001008708.1 | 1, 2 | 240 | ||
| GenBank mRNA | BC017941.1 | - | 1, 2 | 213 | 4282697 | |
| BC019239.1 | - | 1, 2 | 244 | 4050858 | ||
| BC025376.1 | - | 1, 2 | 244 | 4045703 | ||
| BC053896.1 | - | 1, 2 | 217 | 6174351 | ||

CHAC2
Genomic Map