siRNA Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256758.1 | NP_001243687.1 | 1 | 78 | ||
NM_152470.2 | NP_689683.2 | 1 | 78 | |||
NR_046357.1 | 1 | 78 | ||||
XM_011526016.2 | XP_011524318.1 | 1 | 78 | |||
XM_017025788.1 | XP_016881277.1 | 1 | 85 | |||
GenBank mRNA | AK055997.1 | - | 2 | 217 | ||
AK122819.1 | - | 1 | 78 | |||
AK127467.1 | - | 1 | 78 | |||
DA077941.1 | - | 1 | 61 |
