siRNA Details
| Gene Symbol: MS4A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012417.2 | NP_001012417.2 | 6 | 668 | ||
| NM_001100909.1 | NP_001094379.1 | 5 | 548 | |||
| NM_001278320.1 | NP_001265249.1 | 4 | 491 | |||
| GenBank mRNA | AY324188.1 | - | 6 | 668 | ||
| AY324189.1 | - | 4 | 491 | |||
| AY324190.1 | - | 5 | 548 | |||
| BC140799.1 | - | 6 | 542 | 9021667 | ||

MS4A13
Genomic Map