siRNA Details
| Gene Symbol: DNAJC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015882.2 | NP_001015882.2 | 2 | 496 | ||
| NR_037148.1 | 3 | 734 | ||||
| GenBank mRNA | AF447177.1 | - | 3 | 703 | ||
| AK095452.1 | - | 3 | 717 | |||
| BC048318.1 | - | 2 | 467 | 4456137 | ||

DNAJC25
Genomic Map