siRNA Details
| Gene Symbol: ZFAT-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002438.1 | 1 | 229 | |||
| GenBank mRNA | AB167742.1 | - | 1 | 229 | ||
| Gene Symbol: ZFAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB167741.1 | - | 7 | 4710 | ||

ZFAT-AS1
Genomic Map