siRNA Details
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031672.2 | NP_001026842.2 | 4 | 565 | ||
GenBank mRNA | AK090456.1 | - | 4 | 503 | ||
BC064419.1 | - | 4 | 528 | |||
BC071735.1 | - | 4 | 504 | |||
CD515193.1 | - | 2 | 311 |
