siRNA Details
| Gene Symbol: C8orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103552.1 | 1 | 365 | |||
| GenBank mRNA | AK055534.1 | - | 1 | 365 | ||
| BC132945.1 | - | 1 | 234 | 40146967 | ||
| BC136882.1 | - | 1 | 234 | 9020873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8orf49| Gene Symbol: C8orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103552.1 | 1 | 365 | |||
| GenBank mRNA | AK055534.1 | - | 1 | 365 | ||
| BC132945.1 | - | 1 | 234 | 40146967 | ||
| BC136882.1 | - | 1 | 234 | 9020873 | ||