siRNA Details
Gene Symbol: LOC606724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002454.2 | 1 | 438 | |||
GenBank mRNA | U35342.1 | - | 1 | 438 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC606724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002454.2 | 1 | 438 | |||
GenBank mRNA | U35342.1 | - | 1 | 438 |