siRNA Details
Gene Symbol: LOC606724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002454.2 | 1 | 64 | |||
GenBank mRNA | U35342.1 | - | 1 | 64 |
Gene Symbol: CORO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022885.1 | XP_016878374.1 | 10 | 1757 | ||
XM_017022886.1 | XP_016878375.1 | 10 | 1579 | |||
GenBank mRNA | AK097964.1 | - | 2 | 1908 | ||
AK123401.1 | - | 3 | 2563 |
