siRNA Details
| Gene Symbol: LOC613038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002557.1 | 5 | 1458 | |||
| Gene Symbol: LOC388242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002556.1 | 5 | 1458 | |||

LOC613038
Genomic Map