siRNA Details
| Gene Symbol: LOC613038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002557.1 | 2 | 707 | |||
| GenBank mRNA | AW136592.1 | - | 1 | 31 | ||
| Gene Symbol: LOC388242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002556.1 | 2 | 707 | |||

LOC613038
Genomic Map