siRNA Details
| Gene Symbol: EFNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001405.3 | NP_001396.2 | 3 | 522 | ||
| XM_017026449.1 | XP_016881938.1 | 4 | 2327 | |||
| XM_017026450.1 | XP_016881939.1 | 4 | 1211 | |||
| GenBank mRNA | AJ007292.1 | - | 3 | 522 | ||

EFNA2
Genomic Map