siRNA Details
| Gene Symbol: ANP32C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012403.1 | NP_036535.1 | 1 | 260 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANP32C| Gene Symbol: ANP32C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012403.1 | NP_036535.1 | 1 | 260 | ||