siRNA Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281727.1 | NP_001268656.1 | 2, 3 | 154 | ||
| NM_001281728.1 | NP_001268657.1 | 3, 4 | 233 | |||
| NM_001319157.1 | NP_001306086.1 | 2, 3 | 158 | |||
| NM_014183.3 | NP_054902.1 | 2, 3 | 158 | |||
| NM_177953.2 | NP_808852.1 | 3, 4 | 237 | |||
| GenBank mRNA | AF132750.1 | - | 2, 3 | 138 | ||
| AF161511.1 | - | 2, 3 | 129 | |||
| AF165516.1 | - | 2, 3 | 111 | |||
| AF178431.1 | - | 1, 2 | 83 | |||
| AK026864.1 | - | 1, 2 | 94 | |||
| AK294214.1 | - | 2, 3 | 116 | |||
| AY026513.1 | - | 1, 2 | 83 | |||
| BC002481.1 | - | 2, 3 | 136 | 3347555 | ||
| BG110161.1 | - | 2, 3 | 143 | |||
| BM791453.1 | - | 3, 4 | 219 | |||
| BQ639804.1 | - | 3, 4 | 206 | |||
| BU163605.1 | - | 177 | ||||
| CB149456.1 | - | 2, 3 | 154 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025947, NM_001291108, AK003435, CO041603, AK008595, and others.

DYNLRB1
Genomic Map