siRNA Details
| Gene Symbol: TTTY8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001533.1 | 2 | 325 | |||
| Gene Symbol: TTTY8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003591.1 | 2 | 325 | |||
| GenBank mRNA | AA620621.1 | - | 2 | 205 | ||

TTTY8
Genomic Map