siRNA Details
| Gene Symbol: ZNF682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077349.1 | NP_001070817.1 | 4 | 1052 | ||
| NM_033196.2 | NP_149973.1 | 4 | 1095 | |||
| XM_017027455.1 | XP_016882944.1 | 3 | 1199 | |||
| XM_017027456.1 | XP_016882945.1 | 5 | 1159 | |||
| GenBank mRNA | AK027845.1 | - | 2 | 847 | ||
| AK074843.1 | - | 1 | 358 | |||
| AK096571.1 | - | 4 | 1093 | |||
| AK098164.1 | - | 6 | 1298 | |||
| AK127015.1 | - | 1 | 1657 | |||
| BC136304.1 | - | 3 | 1012 | 9020291 | ||

ZNF682
Genomic Map