siRNA Details
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024204.1 | 4 | 623 | |||
NR_024205.1 | 3 | 459 | ||||
NR_024206.1 | 2 | 313 | ||||
GenBank mRNA | AK312010.1 | - | 3 | 372 | ||
BC009508.2 | - | 2 | 278 | |||
BC070202.1 | - | 236 | ||||
BM989746.1 | - | 2 | 308 | |||
CF129206.1 | - | 2 | 351 | |||
DW440910.1 | - | 2 | 330 |
Gene Symbol: MIR4435-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024373.2 | 2 | 399 | |||
GenBank mRNA | BC014776.1 | - | 2 | 326 | ||
BC070201.1 | - | 2 | 286 |
