siRNA Details
| Gene Symbol: LINC00152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024204.1 | 4 | 758 | |||
| NR_024205.1 | 3 | 594 | ||||
| NR_024206.1 | 2 | 448 | ||||
| GenBank mRNA | BC009508.2 | - | 2 | 413 | ||
| BC070202.1 | - | 371 | ||||
| BM989746.1 | - | 2 | 443 | |||
| CF129206.1 | - | 2 | 486 | |||

LINC00152
Genomic Map