siRNA Details
| Gene Symbol: ST13P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002183.1 | 1 | 139 | |||
| GenBank mRNA | AF539468.1 | - | 1 | 139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST13P4| Gene Symbol: ST13P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002183.1 | 1 | 139 | |||
| GenBank mRNA | AF539468.1 | - | 1 | 139 | ||