siRNA Details
| Gene Symbol: CXorf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025265.2 | NP_001020436.1 | 6 | 745 | ||
| NR_033212.1 | 6 | 726 | ||||
| XM_005262244.3 | XP_005262301.1 | 5 | 861 | |||
| GenBank mRNA | AA889552.1 | - | 1 | 39 | ||
| BC144434.1 | - | 6 | 543 | |||

CXorf65
Genomic Map