siRNA Details
| Gene Symbol: ST7-OT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002329.1 | 3 | 470 | |||
| GenBank mRNA | BC043268.2 | - | 3 | 470 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST7-OT4| Gene Symbol: ST7-OT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002329.1 | 3 | 470 | |||
| GenBank mRNA | BC043268.2 | - | 3 | 470 | ||