siRNA Details
Gene Symbol: LCE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178429.3 | NP_848516.1 | 1 | 395 | ||
GenBank mRNA | BC125197.1 | - | 1 | 395 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LCE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178429.3 | NP_848516.1 | 1 | 395 | ||
GenBank mRNA | BC125197.1 | - | 1 | 395 |