siRNA Details
| Gene Symbol: INSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031853.4 | NP_001027024.3 | 9 | 1321 | ||
| NM_001042536.2 | NP_001036001.1 | 9 | 1245 | |||
| NM_001278313.1 | NP_001265242.1 | 8 | 1207 | |||
| NM_001278314.1 | NP_001265243.1 | 9 | 1312 | |||
| NM_001278315.1 | NP_001265244.1 | 8 | 1207 | |||
| NM_001278316.1 | NP_001265245.1 | 7 | 1081 | |||
| XM_006718227.3 | XP_006718290.1 | 10 | 1374 | |||
| XM_011520085.1 | XP_011518387.1 | 9 | 1249 | |||
| XM_011520086.1 | XP_011518388.1 | 9 | 1249 | |||
| XM_017017697.1 | XP_016873186.1 | 10 | 1286 | |||
| XM_017017698.1 | XP_016873187.1 | 9 | 1249 | |||
| GenBank mRNA | AB231745.1 | - | 8 | 1201 | ||
| AB231746.1 | - | 9 | 1306 | |||
| AB231747.1 | - | 7 | 1075 | |||
| AB231748.1 | - | 4 | 576 | |||
| AB236158.1 | - | 9 | 1245 | |||
| AB236159.1 | - | 9 | 1321 | |||
| BC127700.1 | - | 8 | 1207 | 40127962 | ||

INSC
Genomic Map