siRNA Details
| Gene Symbol: SMG1P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135312.1 | 14 | 1895 | |||
| GenBank mRNA | DA126854.1 | - | 3 | 270 | ||
| Gene Symbol: SMG1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096982.1 | - | 8 | 1154 | ||

SMG1P6
Genomic Map