siRNA Details
Gene Symbol: LOC102724699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125409.1 | 2 | 277 | |||
GenBank mRNA | AK094454.1 | - | 2 | 277 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125409.1 | 2 | 277 | |||
GenBank mRNA | AK094454.1 | - | 2 | 277 |