siRNA Details
| Gene Symbol: SPATA31D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145197.1 | NP_001138669.1 | 4 | 423 | ||
| GenBank mRNA | AK125847.1 | - | 4 | 368 | ||
| AK308327.1 | - | 4 | 364 | |||
| Gene Symbol: SPATA31D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207416.2 | NP_997299.2 | 4 | 423 | ||

SPATA31D4
Genomic Map