siRNA Details
| Gene Symbol: LOC401357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130727.1 | 1 | 482 | |||
| GenBank mRNA | AK125790.1 | - | 1 | 482 | ||
| AK128114.1 | - | 1 | 223 | |||
| BC137417.1 | - | 1 | 217 | |||
| JX508854.1 | - | 1 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401357| Gene Symbol: LOC401357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130727.1 | 1 | 482 | |||
| GenBank mRNA | AK125790.1 | - | 1 | 482 | ||
| AK128114.1 | - | 1 | 223 | |||
| BC137417.1 | - | 1 | 217 | |||
| JX508854.1 | - | 1 | 192 | |||