siRNA Details
| Gene Symbol: INTS4P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027392.2 | 5, 6 | 772 | |||
| GenBank mRNA | BC111554.1 | - | 5, 6 | 765 | 40024359 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

INTS4P2| Gene Symbol: INTS4P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027392.2 | 5, 6 | 772 | |||
| GenBank mRNA | BC111554.1 | - | 5, 6 | 765 | 40024359 | |