siRNA Details
| Gene Symbol: PRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005039.3 | NP_005030.2 | 3 | 840 | ||
| NM_199353.2 | NP_955385.1 | 4 | 441 | |||
| NM_199354.2 | NP_955386.1 | 4 | 381 | |||
| GenBank mRNA | BC044827.1 | - | 3 | 826 | ||
| BC141917.1 | - | 4 | 441 | 40113876 | ||
| BG216715.1 | - | 2 | 523 | |||
| K03204.1 | - | 3 | 840 | |||
| K03205.1 | - | 4 | 429 | |||
| K03206.1 | - | 4 | 377 | |||

PRB1
Genomic Map