siRNA Details
| Gene Symbol: LOC102723753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135178.1 | 7 | 1018 | |||
| GenBank mRNA | AK300387.1 | - | 7 | 1018 | ||
| Gene Symbol: HERC2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109773.1 | 7 | 1018 | |||
| GenBank mRNA | BX371795.1 | - | 6 | 836 | ||

LOC102723753
Genomic Map