siRNA Details
Gene Symbol: DPY19L2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036482.1 | 2 | 1010 | |||
GenBank mRNA | BC036244.1 | - | 2 | 1012 | ||
BC047708.1 | - | 2 | 679 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DPY19L2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036482.1 | 2 | 1010 | |||
GenBank mRNA | BC036244.1 | - | 2 | 1012 | ||
BC047708.1 | - | 2 | 679 |