siRNA Details
Gene Symbol: CLN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018941.3 | NP_061764.2 | 3 | 5926 | ||
GenBank mRNA | AK096736.1 | - | 1 | 620 | ||
BC136825.1 | - | 1 | 542 | 9020816 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018941.3 | NP_061764.2 | 3 | 5926 | ||
GenBank mRNA | AK096736.1 | - | 1 | 620 | ||
BC136825.1 | - | 1 | 542 | 9020816 |