siRNA Details
Gene Symbol: TLX1NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130722.1 | 3 | 1554 | |||
NR_130723.1 | 3 | 1525 | ||||
GenBank mRNA | Y11608.1 | - | 1 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TLX1NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130722.1 | 3 | 1554 | |||
NR_130723.1 | 3 | 1525 | ||||
GenBank mRNA | Y11608.1 | - | 1 | 79 |