siRNA Details
| Gene Symbol: TLX1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130722.1 | 3 | 1585 | |||
| NR_130723.1 | 3 | 1556 | ||||
| GenBank mRNA | Y11608.1 | - | 1 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLX1NB| Gene Symbol: TLX1NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130722.1 | 3 | 1585 | |||
| NR_130723.1 | 3 | 1556 | ||||
| GenBank mRNA | Y11608.1 | - | 1 | 110 | ||