siRNA Details
| Gene Symbol: C8orf88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190972.1 | NP_001177901.1 | 5 | 420 | ||
| XM_017012939.1 | XP_016868428.1 | 5 | 346 | |||
| GenBank mRNA | BI562585.1 | - | 5 | 414 | ||
| BI829215.1 | - | 5 | 419 | |||
| DA838443.1 | - | 5 | 383 | |||

C8orf88
Genomic Map