siRNA Details
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054818.1 | - | 1 | 1277 | ||
AK093927.1 | - | 1 | 670 | |||
AK125445.1 | - | 1 | 2124 | |||
AK128507.1 | - | 1 | 3112 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054818.1 | - | 1 | 1277 | ||
AK093927.1 | - | 1 | 670 | |||
AK125445.1 | - | 1 | 2124 | |||
AK128507.1 | - | 1 | 3112 |