siRNA Details
| Gene Symbol: PP13439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF318333.1 | - | 2 | 2216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PP13439| Gene Symbol: PP13439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF318333.1 | - | 2 | 2216 | ||