siRNA Details
| Gene Symbol: ST8SIA6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034129.1 | 3 | 461 | |||
| GenBank mRNA | AK127982.1 | - | 3 | 446 | ||
| AW449205.1 | - | 1 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST8SIA6-AS1| Gene Symbol: ST8SIA6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034129.1 | 3 | 461 | |||
| GenBank mRNA | AK127982.1 | - | 3 | 446 | ||
| AW449205.1 | - | 1 | 110 | |||