siRNA Details
| Gene Symbol: HS6ST2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046691.1 | 2 | 191 | |||
| GenBank mRNA | BQ001311.1 | - | 2 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS6ST2-AS1| Gene Symbol: HS6ST2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046691.1 | 2 | 191 | |||
| GenBank mRNA | BQ001311.1 | - | 2 | 372 | ||