siRNA Details
Gene Symbol: LINC01159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110373.1 | 1 | 131 | |||
NR_110374.1 | 1 | 131 | ||||
GenBank mRNA | DB492871.1 | - | 1 | 134 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110373.1 | 1 | 131 | |||
NR_110374.1 | 1 | 131 | ||||
GenBank mRNA | DB492871.1 | - | 1 | 134 |