siRNA Details
Gene Symbol: CCDC92B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025489.1 | XP_016880978.1 | 2, 3 | 192 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC92B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025489.1 | XP_016880978.1 | 2, 3 | 192 |